EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Synthesis and SAR of tetrahydropyrrolo[1,2-b][1,2,5]thiadiazol-2(3H)-one 1,1-dioxide analogues as highly potent selective androgen receptor modulators.
PMID 17574413
Abstract
Replacement of the 3-oxo group of 2-chloro-4-[(7R,7aS)-7-hydroxy-1,3-dioxotetrahydro-1H-pyrrolo[1,2c]imidazol-2(3H)-yl]-3-methylbenzonitrile resulted in a sulfamide series of selective androgen receptor modulator (SARM) agonists.