EMAIL THIS PAGE TO A FRIEND
Molecular Cancer Therapeutics
Determinants of sensitivity to lovastatin-induced apoptosis in multiple myeloma.
PMID 17575117
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
LP1 VLDL, human