EMAIL THIS PAGE TO A FRIEND
International journal of pancreatology : official journal of the International Association of Pancreatology
Perspectives of CCK antagonists in pancreatic research. Part II. Experimental studies.
PMID 1757727
Abstract
In this article, the effects of different classes of cholecystokinin (CCK) receptor antagonists in CCK-related physiological processes of the pancreas have been discussed. Both glutaramic acid derivatives and natural (benzodiazepine) analogs are potent, competitive antagonists of peripheral CCK receptors. These compounds thus provide a powerful tool for investigating the physiological and pharmacological actions of CCK in the gastrointestinal system, and have already clarified the role of CCK in pancreatic secretion and trophism or growth.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CBR00275
5-Amino-5-oxopentanoic acid, AldrichCPR
C5H9NO3