EMAIL THIS PAGE TO A FRIEND
Nucleosides, nucleotides & nucleic acids
Novel regioselective formation of S- and N-hydroxyl-alkyls of 5-(3-chlorobenzo[b]thien-2-yl)-3-mercapto-4H-1,2,4-triazole and a facile synthesis of triazolo-thiazoles and thiazolo-triazoles. Role of catalyst and microwave.
PMID 17578742
Abstract
Regioselective alkylation of 5-(3-chlorobenzo[b]thien-2-yl)-4H-1,2,4-triazole (1) with hydroxy alkylating agents 2, 3, 13, and the 2,3-O-isopropylidene-1-O-(p-tolylsulfonyl)-glycerol (10) afforded the corresponding S-alkylated derivatives 6, 7, 11, and 14 under both conventional and microwave irradiation conditions; bentonite as a solid support gave better results, with no change in regioselectivity. A facile intramolecular dehydrative ring closure of 6, 7, 11, and 14 using K(2)CO(3) in DMF afforded the corresponding fused triazolo-thiazines and thiazolo-triazole 17-19. The isopropylidenes and acetyl derivatives of the products were prepared.