EMAIL THIS PAGE TO A FRIEND
Macromolecular bioscience
Polyelectrolyte complexes based on (quaternized) poly[(2-dimethylamino)ethyl methacrylate]: behavior in contact with blood.
PMID 17578836
Abstract
Polyelectrolyte complexes (PECs) between (quaternized) poly[2-(dimethylamino)ethyl methacrylate] (PDMAEMA) and (crosslinked) N-carboxyethylchitosan (CECh) or poly(2-acrylamido-2-methylpropane sodium sulfonate) (PAMPSNa) were prepared and characterized in terms of their stability, equilibrium water content, and surface morphology. The evaluation of the behavior of the studied PECs in contact with blood revealed that the (crosslinked) CECh/(quaternized) PDMAEMA complexes had lost the inherent PDMAEMA cytotoxicity but still preserved haemostatic activity. In contrast, the complex formation between (quaternized) PDMAEMA and PAMPSNa allowed the preparation of materials with improved blood compatibility.