EMAIL THIS PAGE TO A FRIEND
Se pu = Chinese journal of chromatography
[Determination of organometallic antitumor crude drug titanocene dichloride by NP-HPLC].
PMID 17580706
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
234826
Bis(cyclopentadienyl)titanium(IV) dichloride, 97%
C10H10Cl2Ti