EMAIL THIS PAGE TO A FRIEND
Se pu = Chinese journal of chromatography
[Determination of cartap residues in tea by GC/micro-ECD].
PMID 17580709
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45995
Cartap hydrochloride, PESTANAL®, analytical standard
C7H15N3O2S2 · HCl