EMAIL THIS PAGE TO A FRIEND
Organic letters
A novel construction of dibenzofuran-1,4-diones by oxidative cyclization of quinone-arenols.
PMID 17580881
Abstract
A novel oxidative cyclization of quinone-arenols 5 leading to products 6 with a dibenzofuran-1,4-dione structure, which forms the core of several natural products, has been developed and applied to the synthesis of violet-quinone (4).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
232017
Tetrachloro-1,4-benzoquinone, 99%
C6Cl4O2