EMAIL THIS PAGE TO A FRIEND
Zhongguo dang dai er ke za zhi = Chinese journal of contemporary pediatrics
[Management of severe fluoroacetamide poisoning with hemoperfusion in children].
PMID 17582269
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
128341
Fluoroacetamide
C2H4FNO