EMAIL THIS PAGE TO A FRIEND
Endocrinology
Activation of estrogen receptor-mediated gene transcription by the equine estrogen metabolite, 4-methoxyequilenin, in human breast cancer cells.
PMID 17584965
Abstract
4-Methoxyequilenin (4-MeOEN) is an O-methylated metabolite in equine estrogen metabolism. O-methylation of catechol estrogens is considered as a protective mechanism; however, comparison of the properties of 4-MeOEN with estradiol (E(2)) in human breast cancer cells showed that 4-MeOEN is a proliferative, estrogenic agent that may contribute to carcinogenesis. 4-MeOEN results from O-methylation of 4-hydroxyequilenin, a major catechol metabolite of the equine estrogens present in hormone replacement therapeutics, which causes DNA damage via quinone formation, raising the possibility of synergistic hormonal and chemical carcinogenesis. 4-MeOEN induced cell proliferation with nanomolar potency and induced estrogen response element (ERE)-mediated gene transcription of an ERE-luciferase reporter and the endogenous estrogen-responsive genes pS2 and TGF-alpha. These estrogenic actions were blocked by the antiestrogen ICI 182,780. In the standard radioligand estrogen receptor (ER) binding assay, 4-MeOEN showed very weak binding. To test for alternate ligand-ER-independent mechanisms, the possibility of aryl hydrocarbon receptor (AhR) binding and ER-AhR cross talk was examined using a xenobiotic response element-luciferase reporter and using AhR small interfering RNA silencing in the ERE-luciferase reporter assay. The results negated the possibility of AhR-mediated estrogenic activity. Comparison of gene transcription time course, ER degradation, and rapid activation of MAPK/ERK in MCF-7 cells demonstrated that the actions of 4-MeOEN mirrored those of E(2) with potency for classical and nonclassical estrogenic pathways bracketing that of E(2). Methylation of 4-OHEN may not represent a detoxification pathway because 4-MeOEN is a full, potent estrogen agonist.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
33693
Equilenin solution, 100 μg/mL in acetonitrile, VETRANAL®, analytical standard
C18H18O2