EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Reactions between Weinreb amides and 2-magnesiated oxazoles: a simple and efficient preparation of 2-acyl oxazoles.
PMID 17585818
Abstract
Treatment of oxazole or 5-aryl oxazoles with i-PrMgCl smoothly generates the corresponding 2-Grignard reagents, which react with Weinreb amides to provide exclusively 2-acyl oxazole products.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
540838
4-Piperidine butyric acid hydrochloride, 97%
C9H18ClNO2
D163708
N,O-Dimethylhydroxylamine hydrochloride, 98%
C2H7NO · HCl