EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Palladium-catalyzed cross-coupling of aryl or vinyl iodides with ethyl diazoacetate.
PMID 17589999
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E22201
Ethyl diazoacetate, contains ≥13 wt. % dichloromethane
C4H6N2O2