EMAIL THIS PAGE TO A FRIEND
Journal of Ethnopharmacology
Dioscorea as the principal herb of Die-Huang-Wan, a widely used herbal mixture in China, for improvement of insulin resistance in fructose-rich chow-fed rats.
PMID 17590296