EMAIL THIS PAGE TO A FRIEND
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Relative importance of mechanisms needs clarification.
PMID 17592156
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A8016
Adenosine 5′-[β-thio]diphosphate trilithium salt, ≥80% (HPLC)
C10H12Li3N5O9P2S