EMAIL THIS PAGE TO A FRIEND
Luminescence : the journal of biological and chemical luminescence
An attempt to design an isoluminol-hydrogen peroxidase-amplified CL that measures intracellularly produced H2O2 in phagocytes: sensitivity for H2O2 is not high enough to allow detection.
PMID 17610299
Abstract
A technique was designed for the determination of hydrogen peroxide release. We found, however, that the isoluminol-amplified chemiluminescence technique is a suitable tool for measuring secretion of superoxide but not hydrogen peroxide.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A8264
4-Aminophthalhydrazide
C8H7N3O2