EMAIL THIS PAGE TO A FRIEND
Skin research and technology : official journal of International Society for Bioengineering and the Skin (ISBS) [and] International Society for Digital Imaging of Skin (ISDIS) [and] International Society for Skin Imaging (ISSI)
Influence of skin colour on the detection of cutaneous erythema and tanning phenomena using reflectance spectrophotometry.
PMID 17610644
Abstract
This research aims at assessing the influence of baseline skin colour on the ability of reflectance spectrophotometry to detect cutaneous erythema induced by a low concentration of methyl nicotinate (2.5 mM) (first objective), and to detect tanning induced by ultraviolet rays (UVA+UVB) at infra-erythemal doses (second objective). Two independent studies were conducted to reach their respective objectives, on 27 women for the first study and on 12 women for the second study. Skin colour measurements were expressed in two different ways: percentages of reflected light at increasing wavelengths lambda (400 nm or =40 degrees. The assumption is that in the darkest skins, the emitted light is mainly absorbed by the melanin in the epidermis. Otherwise, after UV irradiation, the tanning was detectable only for individuals with fair to dark skin defined by ITA <50 degrees. This can be explained by the fact that UV stimulation of the fairest skin subjects, known to be melano-compromised individuals, can only produce a weak tanning that our study did not succeed in detecting.