EMAIL THIS PAGE TO A FRIEND
Experimental parasitology
Antiplasmodial structure-activity relationship of 3-trifluoromethyl-2-arylcarbonylquinoxaline 1,4-di-N-oxide derivatives.
PMID 17612525
Abstract
Derivatives of 3-trifluoromethyl-2-arylcarbonylquinoxaline 1,4-di-N-oxide (4b-g, 5b-g, 6a-g) were synthesized and evaluated for their capacity to inhibit the growth of chloroquine-resistant Plasmodium falciparum FCB1 strain in culture. Compound 7-chloro-2-(2-furylcarbonyl)-3-trifluoromethyl-1,4-quinoxaline di-N-oxide (5g) was the most active being almost 5 times more active than chloroquine. It was also 50 times more active against P. falciparum than toxic toward MCF7 cells. Structural characteristics for a quinoxaline to be active were defined: bioisosteric modification of phenyl group by 2-thienyl or 2-furyl subunits, R2 position must be free or occupied by a methyl group and R1 position must be free or occupied by Cl, CH3, OCH3 or CF3.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
426016
4,4,4-Trifluoro-1-(2-furyl)-1,3-butanedione, 99%
C8H5F3O3