EMAIL THIS PAGE TO A FRIEND
Histology and histopathology
Role of fibrosis-related genes and pancreatic duct obstruction in rat pancreatitis models: implications for chronic pancreatitis.
PMID 17616939
Abstract
Human chronic pancreatitis is characterized by irreversible fibrosis, whereas pancreatic fibrosis in animal models is reversible. In this study, we compare the development of pancreatic fibrosis in the dibutyltin dichloride (DBTC) model, WBN/Kob rats and bile duct-ligated (BDL) rats. DBTC (8 mg/kg) was administered to LEW rats, and the pancreas was histopathologically investigated sequentially. Male and female WBN/Kob rats aged 4, 6 and 8 months were also examined. BDL rats were prepared by ligation of the bile duct at the duodenal portion and sacrificed at 3 or 7 days after ligation. Fibrosis in the DBTC model peaked after 1 week and was limited to the areas around the pancreatic ducts after 2 weeks, and was composed of both type I and type III collagen. In contrast, fibrosis in male WBN/Kob rats peaked at age 4 months, expanded into intralobular area, and was composed of type III collagen. It exhibited almost no type I collagen and a marked tendency to regress. Pancreatic fibrosis in BDL rats was somewhat difficult to induce and required increased stimulation. This suggests that fibrosis in human biliary pancreatitis may gradually form based on weak, continuous stimulation. We conclude that type I collagen may be involved in the progression of irreversible fibrosis. The imbalance between synthesis and degradation of extracellular matrix molecules or degree of stimulation over a certain period may lead to pancreatic fibrosis. Gene expressions of prolyl hydroxylase and tissue inhibitors of matrix metalloproteinase-2 were elevated.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
205494
Dibutyltin dichloride, 96%
C8H18Cl2Sn