EMAIL THIS PAGE TO A FRIEND
Bioscience, biotechnology, and biochemistry
Effects of the amino-carbonyl reaction of ribose and glycine on the formation of the 2(or 5)-ethyl-5(or 2)-methyl-4-hydroxy-3(2H)-furanone aroma component specific to miso by halo-tolerant yeast.
PMID 17617718
Abstract
The formation of HEMF[2(or 5)-ethyl-5(or 2)-methyl-4-hydroxy-3(2H)-furanone], the aroma component specific to miso and soy sauce, was promoted by cultivating the halo-tolerant yeast, Zygosaccharomyces rouxii, in a medium including the amino-carbonyl reaction products based on ribose and glycine. The glucose concentration in the medium influenced the HEMF formation by Z. rouxii.
Related Materials