EMAIL THIS PAGE TO A FRIEND
MMW Fortschritte der Medizin
[Aso an inspection of the family dog is warrented here].
PMID 17621788
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D2205000 Dimetindene maleate, European Pharmacopoeia (EP) Reference Standard
C20H24N2 · C4H4O4