EMAIL THIS PAGE TO A FRIEND
Applied biochemistry and biotechnology
Reusability of entrapped cells of Pseudomonas diminuta for production of 7-aminocephalosporanic acid.
PMID 17625270
Abstract
Entrapped cells of P. diminuta were used for the production of 7-aminocephalosporanic acid (7-ACA), a key intermediate required for the production of most of the clinically used cephalosporin derivatives, i.e., semisynthetic cephalosporins. The repeated batch production of 7-ACA with entrapped cells of P. diminuta in different carriers were carried out for six cycles at optimal conditions. It was found that 33% , 38%, and 47% of activity was lost with chitosan, gelatin, and agar, respectively as immobilizing supports after the sixth cycle of operation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
191140
7-Aminocephalosporanic acid, 98%
C10H12N2O5S