EMAIL THIS PAGE TO A FRIEND
Journal of virology
Polyomavirus small T antigen controls viral chromatin modifications through effects on kinetics of virus growth and cell cycle progression.
PMID 17626093
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
06-911
Anti-acetyl-Histone H3 (Lys14) Antibody, Use Anti-acetyl-Histone H3 (Lys14) Antibody (Rabbit Polyclonal Antibody) validated in ICC, IP, WB to detect acetyl-Histone H3 (Lys14) also known as H3K14Ac, Histone H3 (acetyl K14).
06-942
Anti-acetyl-Histone H3 (Lys9) Antibody, Anti-acetyl-Histone H3 (Lys9) Antibody is a Rabbit Polyclonal Antibody for detection of Histone H3 acetylated on lysine 9. This purified Ab, also known as Anti-H3K9Ac, is specificity verified by dot blot (DB), published in peer reviewed journals, and validated in WB, ChIP, ChIP-seq, FC.
06-599
Anti-acetyl-Histone H3 Antibody, Detect acetyl-Histone H3 with Anti-acetyl-Histone H3 Antibody (Rabbit Polyclonal Antibody), that has been shown to work in WB, ICC, ChIP, ChIP-seq.
07-327
Anti-acetyl-Histone H4 (Lys5) Antibody, Use Anti-acetyl-Histone H4 (Lys5) Antibody (Rabbit Polyclonal Antibody) validated in WB, DB, ICC, ChIP-seq, ChIP to detect acetyl-Histone H4 (Lys5) also known as H4K5Ac, Histone H4 (acetyl K5).
06-598
Anti-acetyl-Histone H4 Antibody, Anti-acetyl-Histone H4 Antibody is a Rabbit Polyclonal Antibody for detection of acetyl-Histone H4 also known as H4 histone family member A, histone 1-H4a & has been validated in ChIP, ICC, IP & WB.
07-030
Anti-dimethyl-Histone H3 (Lys4) Antibody, Use Anti-dimethyl-Histone H3 (Lys4) Antibody (rabbit polyclonal antibody) is published and validated in ChIP, DB, ICC, IF, WB, PIA, ChIP-seq. to detect dimethyl-Histone H3 (Lys4) also known as H3K4me2, Histone H3 (di methyl K4).
05-499
Anti-Histone H3 Antibody, clone 6.6.2, Anti-Histone H3 Antibody, clone 6.6.2 is a Mouse Monoclonal Antibody for detection of Histone H3 also known as H3 histone family 3A & has been validated in WB.
05-858
Anti-Histone H4 Antibody, pan, clone 62-141-13, rabbit monoclonal, Anti-Histone H4 Antibody, pan, clone 62-141-13 is a Rabbit Monoclonal Antibody for detection of Histone H4 also known as Histone H4 & has been validated in ChIP, WB, Mplex, ChIP-seq.
06-570
Anti-phospho-Histone H3 (Ser10) Antibody, Mitosis Marker, Anti-phospho-Histone H3 (Ser10) Antibody, Mitosis Marker is a Rabbit Polyclonal Antibody for detection of Histone H3 phosphorylated at serine 10.This highly published Ab, also known as Anti-H3S10p, has been validated in ICC, IP & WB.
07-442
Anti-trimethyl-Histone H3 (Lys9) Antibody, Use Anti-trimethyl-Histone H3 (Lys9) Antibody (rabbit polyclonal antibody) validated in DB, ICC, Mplex, PIA, WB, ChIP-seq to detect trimethyl-Histone H3 (Lys9) also known as H3K9me3, Histone H3 (tri methyl K9).