EMAIL THIS PAGE TO A FRIEND
Carbohydrate research
First synthesis of tepidopterin [2'-O-(2-acetamido-2-deoxy-beta-d-glucopyranosyl)-L-threo-biopterin].
PMID 17631283
Abstract
N(2)-(N,N-Dimethylaminomethylene)-1'-O-(4-methoxybenzyl)-3-[2-(4-nitrophenyl)ethyl]-L-threo-biopterin (14) was prepared from L-xylose in an 11-step-sequence. The first synthesis of tepidopterin (3) was achieved by treatment of 14 with 3,4,6-tri-O-acetyl-2-deoxy-2-phthalimido-beta-D-glucopyranosyl bromide in the presence of silver triflate and tetramethylurea, followed by removal of the protecting groups.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T24503
Tetramethylurea, 99%
C5H12N2O