EMAIL THIS PAGE TO A FRIEND
Journal of clinical psychopharmacology
The association between HTR2C gene polymorphisms and the metabolic syndrome in patients with schizophrenia.
PMID 17632216
Abstract
The use of antipsychotics is associated with metabolic side effects, which put patients with schizophrenia or related disorders at risk for cardiovascular morbidity. The high interindividual variability in antipsychotic-induced metabolic abnormalities suggests that genetic makeup is a possible determinant. In this cross-sectional study, we investigated whether genotypes of the HTR2C receptor are associated with the metabolic syndrome in patients using antipsychotics. Patients were identified from a schizophrenia disease management program. In this program, patients' blood pressure, triglycerides, high-density lipoprotein-cholesterol, and waist circumference are measured regularly during follow-up. The primary end point of our study was the prevalence of the metabolic syndrome as classified by a modified version of the National Cholesterol Education Program's Adult Treatment Panel III. Primary determinants were polymorphisms in the HTR2C receptor gene (HTR2C:c.1-142948[GT]n, rs3813928 [-997 G/A], rs3813929 [-759 C/T], rs518147 [-697 G/C], and rs1414334 [C > G]). The included patients (n = 112) mainly (>80%) used atypical antipsychotics (clozapine, olanzapine, and risperidone). Carriership of the variant alleles of the HTR2C polymorphisms rs518147, rs1414334, and HTR2C:c.1-142948(GT)n was associated with an increased risk of the metabolic syndrome (adjusted odds ratio [OR], 2.62 [95% confidence interval {CI}, 1.00-6.85]; OR, 4.09 [95% CI, 1.41-11.89]; and OR, 3.12 [95% CI, 1.13-8.16]), respectively. Our findings suggest that HTR2C genotypes are associated with antincreased risk of metabolic syndrome in patients taking antipsychotics.