EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
Cloning, nucleotide sequencing, and analysis of the AcrAB-TolC efflux pump of Enterobacter cloacae and determination of its involvement in antibiotic resistance in a clinical isolate.
PMID 17638702
Abstract
Enterobacter cloacae is an emerging clinical pathogen that may be responsible for nosocomial infections. Management of these infections is often difficult, owing to the high frequency of strains that are resistant to disinfectants and antimicrobial agents in the clinical setting. Multidrug efflux pumps, especially those belonging to the resistance-nodulation-division family, play a major role as a mechanism of antimicrobial resistance in gram-negative pathogens. In the present study, we cloned and sequenced the genes encoding an AcrAcB-TolC-like efflux pump from an E. cloacae clinical isolate (isolate EcDC64) showing a broad antibiotic resistance profile. Sequence analysis showed that the acrR, acrA, acrB, and tolC genes encode proteins that display 79.8%, 84%, 88%, and 82% amino acid identities with the respective homologues of Enterobacter aerogenes and are arranged in a similar pattern. Deletion of the acrA gene to yield an AcrA-deficient EcDC64 mutant (EcDeltaacrA) showed the involvement of AcrAB-TolC in multidrug resistance in E. cloacae. However, experiments with an efflux pump inhibitor suggested that additional efflux systems also play a role in antibiotic resistance. Investigation of several unrelated isolates of E. cloacae by PCR analysis revealed that the AcrAB system is apparently ubiquitous in this species.