EMAIL THIS PAGE TO A FRIEND
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica
[Studies on flavones in of Lavandula augustifolia].
PMID 17639984
Abstract
To study on the chemical constituents of Lavandula augustifolia. The compounds were extracted with 95% ethyl alcohol, isolated by various column chromatography and identified by spectroscopic methods. Seven flavanoids were isolated and identified as apigenin (1), ladanein (2), apigenin-7-O-beta-D-(6'-p-hydoxy-cinnamoyloxy) -mannoside (3), luteolin (4), apigenin-7-O-beta-D-glucoside (5), luteolin-7-O-beta-D-glucoside (6), 5, 4'-dihydroxy flavonoid-7-O-beta-D-pyranglycuronate buthyl ester (7). All of these seven compounds were obtained from this plant for the first time.