EMAIL THIS PAGE TO A FRIEND
Neurobiology of aging
Protection against beta-amyloid induced abnormal synaptic function and cell death by Ginkgolide J.
PMID 17640772
Abstract
A new Ginkgo biloba extract P8A (TTL), 70% enriched with terpene trilactones, prevents A beta(1-42) induced inhibition of long-term potentiation in the CA1 region of mouse hippocampal slices. This neuroprotective effect is attributed in large part to ginkgolide J that completely replicates the effect of the extract. Ginkgolide J is also capable of inhibiting cell death of rodent hippocampal neurons caused by A beta(1-42). This beneficial and multi-faceted mode of action of the ginkgolide makes it a new and promising lead in designing therapies against Alzheimer's disease.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
89556
Ginkgolide J, analytical standard
C20H24O10