EMAIL THIS PAGE TO A FRIEND
Nature protocols
Automated generation of a dihydropyrimidine compound library using microwave-assisted processing.
PMID 17641636
Abstract
Here we report the generation of a small focused library of 12 diversely functionalized dihydropyrimidine (DHPM) derivatives via one-pot three-component Biginelli cyclocondensation of beta-ketoesters, aldehydes and (thio)ureas. By applying controlled microwave heating under sealed vessel conditions using a fully automated microwave instrument including a gripper and liquid handler, the sequential synthesis of DHPMs can be performed in a shorter reaction time (10-20 min per one DHPM derivative) compared to conventional heating methods, which normally require several hours of reflux heating. The solid products either crystallize directly upon cooling or can be precipitated upon addition of water, requiring only filtration for isolation. In this way, the DHPM derivatives are obtained in high purity and no further purification by recrystallization or chromatography is necessary. This can be ascribed to the microwave heating technology where less side-product formation is often seen. The preparation of this 12-membered DHPM library can be carried out within approximately 9 h.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
124974
2-Chlorobenzaldehyde, 99%
C7H5ClO
281506
Methyl 4-methoxyacetoacetate, 97%
C6H10O4