EMAIL THIS PAGE TO A FRIEND
Bollettino chimico farmaceutico
3-Aminorhodanine and 2-hydrazino-2-imidazo-line hydrazones. Synthesis and antiparasitic pharmacology.
PMID 1764238
Abstract
Two series of hydrazones were prepared by condensation of various aldehydes with 2-aminorhodanine (series 1) and 2-hydrazino-2-imidazoline (series 2). These compounds showed interesting antiparasitic activities against amoeba, trichomonas, leishmania and plasmodium.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A79507
3-Aminorhodanine, 99%
C3H4N2OS2