EMAIL THIS PAGE TO A FRIEND
Journal of emergency nursing: JEN : official publication of the Emergency Department Nurses Association
Ethylene glycol poisoning: current diagnostic and management issues.
PMID 17643796