EMAIL THIS PAGE TO A FRIEND
Journal of hazardous materials
Simultaneous square wave voltammetric determination of 2-mercaptobenzothiazole and 2-mercaptobenzoxazole by partial least squares method in water samples.
PMID 17651895
Abstract
A rapid sensitive and versatile method for simultaneous determination of 2-mercaptobenzothiazole (MBT) and 2-mercaptobezoxazole (MBO) based on the square wave voltammetric (SWV) using mercury drop electrode (SMDE) has been presented. A three-electrode system containing SMDE working electrode, Pt auxiliary electrode and Ag/AgCl reference electrode was used throughout. The linear calibration graphs are in the concentration range of 7-40 microg mL(-1) and the equations are: (Deltai)=1.372CMBT-9.112 (r=0.9982) and (Deltai)=0.246CMBO-1.736 (r=0.9985) for MBT and MBO, respectively. Partial least squares regression (PLS) was applied to resolve the seriously overlapped voltammograms without any pre-separation step. The five level partial factorial design was used as calibration design method and the cross-validation method was used to select the number of significant factor for PLS model building. Five significant PLS components are used for MBT and MBO. A set of synthetic sample mixtures were used to validate the propose method. The root-mean-square errors of predictions (RMSEPs) and percent of relative prediction errors (RPEs) are 0.841 and 0.777 microg mL(-1) and +/-3.58 and +/-3.74% for MBT and MBO, respectively. The developed method was then applied to the analysis of these two compounds in different water samples with satisfactory results.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M3302
2-Mercaptobenzothiazole, 97%
C7H5NS2
63730
2-Mercaptobenzothiazole, technical, ≥90% (T)
C7H5NS2