EMAIL THIS PAGE TO A FRIEND
Journal of virology
Evidence for involvement of BH3-only proapoptotic members in adenovirus-induced apoptosis.
PMID 17652400
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
06-536
Anti-Bak Antibody, NT, clone, Upstate®, from rabbit