EMAIL THIS PAGE TO A FRIEND
Scandinavian journal of infectious diseases
Successful treatment of ctx-m ESBL producing Escherichia coli relapsing pyelonephritis with long term pivmecillinam.
PMID 17654359
Abstract
Oral therapy options for pyelonephritis caused by ESBL producing E. coli are limited. We describe a woman with relapsing pyelonephritis due to a CTX-M ESBL E. coli who was cured with a prolonged course of pivmecillinam. This suggests pivmecillinam may be effective treatment for selected patients with pyelonephritis with these organisms.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0817
Pivmecillinam, ≥98% (HPLC)
C21H33N3O5S