EMAIL THIS PAGE TO A FRIEND
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy
Crystal structure and photoluminescence of 7-(N,N'-diethylamino)-coumarin-3-carboxylic acid.
PMID 17656149
Abstract
The structure of 7-(N,N'-diethylamino)-coumarin-3-carboxylic acid (DCCA) was verified by single crystal X-ray crystallography. The UV-vis absorption and fluorescence of DCCA were discussed. The compound exhibits strong blue emission under ultraviolet light excitation. The molecular structure of DCCA was optimized using density function theory (DFT) at B3LYP/6-31G level, showing that the optimized geometers parameters are in good agreement with experiment data. In addition, the HOMO and LUMO levels of DCCA were deduced.
Related Materials