EMAIL THIS PAGE TO A FRIEND
Journal of occupational medicine and toxicology (London, England)
Urinary N-acetyl-beta -D-glucosaminidase and its isoenzymes A & B in workers exposed to cadmium at cadmium plating.
PMID 17659077
Abstract
The present study was carried out to determine the effect of cadmium exposure on Urinary N-acetyl-beta -D-glucosaminidase and its isoenzymes A and B in workers exposed at cadmium plating. 50 subjects using cadmium during cadmium plating formed the study group. An equal number of age-sex matched subjects working in administrative section formed the control group. Urinary cadmium levels were determined by using a flameless atomic absorption spectrophotometer. Urinary N-acetyl-beta -D-glucosaminidase and its isoenzymes A and B were determined by using spectrophotmetric method. A significant increase of urinary total N-acetyl-beta -D-glucosaminidase and its isoenzymes A and B profiles were noted in study as compared to controls. The levels of urinary N-acetyl-beta -D-glucosaminidase and its isoenzymes A and B profiles were positively and significantly correlated with cadmium levels in urine. Multiple regression analysis was used to assess the effect of urinary cadmium or life style confounding factors (age, BMI, smoking and alcohol consumption) on urinary N-acetyl-beta -D-glucosaminidase and its isoenzymes A and B. The analysis showed that the study subjects who had urine cadmium levels greater than 5 mug/g of creatinine, work duration >15 years, smoking and body mass index variables were significantly associated with urinary total N-acetyl-beta -D-glucosaminidase but not on isoenzymes A&B. The results presented in this study shows that the increased levels of urinary N-acetyl-beta -D-glucosaminidase observed in cadmium-exposed workers could be used as biomarkers for suggesting preventive measure.