EMAIL THIS PAGE TO A FRIEND
Revista clinica espanola
[Decrease of high density lipoproteins secondary to treatment with anabolizing steroids].
PMID 17662209
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O0380
Oxymetholone, analytical standard
C21H32O3