EMAIL THIS PAGE TO A FRIEND
International journal of clinical practice. Supplement
Pulmonary hypertension and the serotonin hypothesis: where are we now?
PMID 17663674
Abstract
In the 1960s, serotonin (5HT) was associated with pulmonary arterial hypertension (PAH) caused by certain diet pills, but has recently been the subject of renewed interest in the field of PAH. Serotonin can be synthesised in the pulmonary endothelium with the rate-limiting step being the activity of tryptophan hydroxylase1 (Tph1). The serotonin is released and can then: (i) pass into the underlying pulmonary smooth muscle cells through the serotonin transporter (SERT) to initiate proliferation and/or (ii) activate serotonin receptors on pulmonary smooth muscle cells to evoke proliferation and/or contraction. Serotonin may also mediate pulmonary fibroblast proliferation via the SERT and/or serotonin receptors. Here we will unravel, discuss and update the 'serotonin hypothesis' of PAH in light of recent advances in the field. In conclusion, the activity of serotonin receptors, the SERT and Tph1 can all be elevated in clinical and experimental PAH and each offers a potentially unique therapeutic target.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F112
(+)-Fenfluramine hydrochloride
C12H16F3N · HCl