EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Using acetate anions to induce translational isomerization in a neutral urea-based molecular switch.
PMID 17665407
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
379336
Dimethyl 2,6-pyridinedicarboxylate, 99%
C9H9NO4