EMAIL THIS PAGE TO A FRIEND
Acta poloniae pharmaceutica
GC method for quantitative determination of residual 2-(2-chloroethoxy) ethanol (CEE) and N-methyl-2-pyrrolidinone (NMP) in quetiapine.
PMID 17665871
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
162965
2-(2-Chloroethoxy)ethanol, 99%
C4H9ClO2