EMAIL THIS PAGE TO A FRIEND
Natural product reports
The lycopodium alkaloids.
PMID 1766595
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
19108 Lycopodium