EMAIL THIS PAGE TO A FRIEND
Carbohydrate research
An investigation of the inclusion complex of beta-cyclodextrin with p-nitrobenzoic acid in the solid state.
PMID 17669379
Abstract
The weak inclusion complex of cyclomaltoheptaose (beta-cyclodextrin, betaCD) with p-nitrobenzoic acid was investigated in the solid state. Crystallography shows that two betaCD molecules co-crystallize with two p-nitrobenzoic acids and 28.5 water molecules [2(C(42)H(70)O(35))x2(C(7)H(5)NO(4))x28.5H(2)O] in the triclinic system.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
72910
4-Nitrobenzoic acid, purum, ≥98.0% (HPLC)
C7H5NO4
461091
4-Nitrobenzoic acid, 98%
C7H5NO4