EMAIL THIS PAGE TO A FRIEND
The Journal of pharmacy and pharmacology
Derivatization to stabilize some aliphatic primary hydroxylamines for g.l.c. analysis.
PMID 17670
Abstract
By appropriate choice of trimethylsilylating and trifluoroacetylating reagents and organic solvents for extraction, stable derivative of aliphatic primary hydroxylamines metabolites, N-hydroxyphentermine, N-hydroxychlorphentermine, N-hydroxymexiletene, N-hydroxyphenethylamine, N-hydroxyamphetamine, and N-hydroxy-3,4-dimethoxyamphetamine, were obtained and examined by g.l.c. analysis without decomposition and without interference from the parent drug or other metabolic products.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
91719
Trifluoroacetic anhydride, for GC derivatization, ≥99.0% (GC)
C4F6O3