EMAIL THIS PAGE TO A FRIEND
General and comparative endocrinology
Distinct features of dehydrocorticosterone reduction into corticosterone in the liver and duodenum of the domestic fowl (Gallus gallus domesticus).
PMID 17673214
Abstract
The mammalian 11-beta hydroxysteroid dehydrogenase type 1 (11 betaHSD1) reduces glucocorticoids (GC) at C11 from the 11-keto-GC nonactive form to the 11-hydroxy-GC active form, an action essential for survival. Whereas GC metabolism at C11 and the role of 11 betaHSD1 are studied extensively in mammals, information about these in birds is scattered. Herein, we report the GC bidirectional metabolism in chickens. In hens' liver and duodenal mucosa, 11 betaHSD1-like mRNA expression was detected; and 11 betaHSD1-like immunoreactivity was found linked to membranes of hepatocytes and duodenal enterocytes. With either NADH or NADPH, the membranal fraction of liver and duodenal mucosa converted dehydrocorticosterone (A) into corticosterone (B) with K(m) (1.1-8.7 microM) and V(max) (10-40 pmol/mg protein/min) values similar to those reported for mammalian 11 betaHSD1. In the presence of NADP(+) or NAD(+), these membranal fractions oxidized B into A. With either NADPH or NADH, the cytosol of chicken liver and duodenal mucosa reduced A into B (K(m) of 1.1 - 2.3 microM and V(max) of 260-960 pmol/mg protein/min). These cytosolic fractions did not convert any amount of B into A when incubated with either NADP(+) or NAD(+). This may suggest that chicken liver and duodenal mucosa express 11 betaHSD1 that is a membrane-bound oxoreductase which uses both NADPH/NADP(+) and NADH/NAD(+) as cosubstrates. The substantial reduction of A into B (but no conversion of B into A) found in the cytosol is most likely executed by a unidirectional soluble reductase, different than 11 betaHSD1.