EMAIL THIS PAGE TO A FRIEND
Klinicheskaia laboratornaia diagnostika
[Pyrogallol red technique is an alternative to routine urinary protein-determining methods].
PMID 17682474
Abstract
The paper presents the basic urinary protein-determining methods, such as turbidimetric and colorimetric ones and provides their brief analytical characterization. The authors consider the sulfosalicylic technique as most universally employed by clinical diagnostic laboratories and the pyrogallolic one as most promising. The comparative characteristics of both techniques are described and their advantages and disadvantages are noted. In terms of correct estimation and reproducibility of the results of an analysis, the pyrogallic technique is superior to the sulfosalicylic and other methods for protein determination; it can identify microproteinuria and thus a better alternative to the routine methods for urinary protein determination.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
06931
Pyrogallol, analytical standard
C6H6O3
P8759
Pyrogallol Red
C19H12O8S