EMAIL THIS PAGE TO A FRIEND
International MS journal
Treating fatigue.
PMID 17686346
Abstract
Fatigue is recognized as one of the most disabling and common symptoms of MS. However, no universal consensus has been reached regarding a formal definition for fatigue, and its mechanisms and causes are not understood. This review summarizes important research in this field and recommends a multidisciplinary approach to managing MS patients with fatigue. Such an approach should include proven pharmaceutical and non-pharmaceutical treatments.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P0048
Pemoline, ≥98% (HPLC)
C9H8N2O2