EMAIL THIS PAGE TO A FRIEND
Journal of combinatorial chemistry
Regiospecific solid-phase strategy to N7-substituted purines and its application to 8-azapurines and [I]-condensed purines.
PMID 17688325
Abstract
A highly regioselective and traceless solid-phase route to 1,7,8-trisubstituted purines has been developed. This methodology could be extended to the preparation of 8-azapurines and [i]-condensed purines. A representative set of 17 purines, azapurines, and [i]-condensed purines was synthesized. This paper also describes a mild method to prepare the p-benzyloxybenzylamine (BOBA) resin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B22984
O-Benzylhydroxylamine hydrochloride, 99%
C7H9NO · HCl