EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Synthesis and antibacterial activity of some novel chiral fluorophoric biscyclic macrocycles.
PMID 17689076
Abstract
Synthesis of chiral permanent fluorophoric biscyclic macrocycles incorporating anthraquinone and (S)-BINOL core is described. Interestingly, the biscyclic macrocycle 1 exhibited remarkable antibacterial activity against most of the pathogenic bacteria in the tested concentrations as compared to the other three compounds 2, 14 and 17 as well as the test control, tetracycline. Further biscyclophanes 1 and 2 exhibited permanent fluorescence sensing property even under highly acidic conditions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
367834
1,4-Diaminoanthraquinone, technical grade, 90%
C14H10N2O2