EMAIL THIS PAGE TO A FRIEND
European journal of pharmacology
Lu 35-138 ((+)-(S)-3-{1-[2-(1-acetyl-2,3-dihydro-1H-indol-3-yl)ethyl]-3,6-dihydro-2H-pyridin-4-yl}-6-chloro-1H-indole), a dopamine D4 receptor antagonist and serotonin reuptake inhibitor: characterisation of its in vitro profile and pre-clinical antipsychotic potential.
PMID 17689529
Abstract
The present study describes the pharmacological profile of the putative antipsychotic drug Lu 35-138 ((+)-(S)-3-{1-[2-(1-acetyl-2,3-dihydro-1H-indol-3-yl)ethyl]-3,6-dihydro-2H-pyridin-4-yl}-6-chloro-1H-indole). The in vitro receptor profile of Lu 35-138 revealed high affinity (K(i)=5 nM) and competitive antagonism (K(b)=8 nM) at dopamine D(4) receptors combined with potent 5-HT uptake inhibition (IC(50)=3.2 nM) and moderate alpha(1)-adrenoceptor affinity (K(i)=45 nM). In vivo, Lu 35-138 selectively counteracted hyperlocomotion induced by d-amphetamine (0.5 mg/kg; ED(50)=4.0 mg/kg, s.c.) in rats and phencyclidine (PCP; 2.5 mg/kg; ED(50)=13 mg/kg, s.c.) in mice. Lu 35-138 was unable to affect hyperlocomotion induced by a high dose of d-amphetamine (2.0 mg/kg), which indicates a preferential action on limbic versus striatal structures. A similar limbic selectivity of Lu 35-138 was indicated in voltammetric measure of dopamine output in the core and shell subdivisions of the nucleus accumbens in rats. Furthermore, a relatively large dose of Lu 35-138 (18 mg/kg, s.c.) counteracted d-amphetamine-induced disruption of pre-pulse inhibition in rats and repeated administration of Lu 35-138 (0.31 or 1.25 mg/kg, p.o. once daily for 3 weeks) reduced the number of spontaneously active dopamine neurones in the ventral tegmental area, underlining its antipsychotic-like profile. Lu 35-138 failed to induce catalepsy in rats or dystonia in Cebus apella monkeys and did not deteriorate spatial memory in rats as assessed by water maze performance. Collectively, these results suggest that Lu 35-138 possesses antipsychotic activity combined with a low extrapyramidal and cognitive side effect liability.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PZ0174
Sonepiprazole hydrate, ≥98% (HPLC)
C21H27N3O3S · xH2O