EMAIL THIS PAGE TO A FRIEND
Chemosphere
Sorption of 1-hydroxy-2-naphthoic acid to goethite, lepidocrocite and ferrihydrite: batch experiments and infrared study.
PMID 17689586
Abstract
The adsorption of naphthoic acids to iron oxides and hydroxides influences strongly their mobility in soils and sediments. Sorption of 1-hydroxy-2-naphthoic acid (HNA) to three iron oxides was examined over a wide range of conditions (pH, ionic strength, sorbate and sorbent concentrations). In the examination of HNA sorption, Tempkin model was performed to fit sorption data of HNA onto all iron oxides. The adsorption in the Henry law range increases in the order: goethite
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
109630
1-Hydroxy-2-naphthoic acid, ≥97.0%
C11H8O3