EMAIL THIS PAGE TO A FRIEND
Cancer chemotherapy and pharmacology
Disruption of Golgi processing by 2-phenyl benzimidazole analogs blocks cell proliferation and slows tumor growth.
PMID 17690881
Abstract
Cancer chemotherapy continues to be challenged by the emergence of resistant tumors, and one organelle entwined in the development of drug resistance is the Golgi apparatus. Recently, we discovered a group of 2-(substituted phenyl)-benzimidazole (2-PB) compounds that displace resident Golgi proteins from the juxtanuclear region resulting in their degradation. These compounds are also potent anti-proliferative agents, which together with their action on the Golgi made a compelling case for testing them against cancer. The anti-tumor activity of a group of 2-PB compounds was examined both in vitro and in vivo. The role of the Golgi in the anti-proliferative effect was assessed by comparing the proliferation of individual cell lines with the distribution and total cellular expression of selected resident Golgi proteins. The anti-proliferative activity of 2-PB compounds is partially reversible (time- and concentration-dependent), non-cell-cycle-specific, and translates to tumor growth inhibition in vivo. While 2-PB compounds displace resident Golgi proteins from the juxtanuclear region in all cells, those that are resistant to the anti-proliferative effects differ from sensitive cells in that they have the capacity to protect these Golgi proteins from degradation. These results illustrate the utility of targeting the Golgi for cancer drug development. They also reveal a cellular strategy for resisting 2-PB drug effects through protection of displaced Golgi proteins from degradation thus allowing their continued function.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P19809
2-Phenylbenzimidazole, 97%
C13H10N2